• USA Home
  • Z111856 - Hydrion® MicroFine pH test paper set
EMAIL THIS PAGE TO A FRIEND
Z111856 Aldrich
Hydrion® MicroFine pH test paper set
12 double-rolls of test papers
Purchase
Description
General description
12 double-rolls of test papers. Includes Hydrion Vivid 9 to 13, which provides unusual accuracy (0.5 pH units) in the difficult, strongly alkaline range and Hydrion Mikro Ultrafast 0 to 14, a combination of Insta-Chek 0 to 13 and the ultra-sharp 12 to 14 pH range paper. Remaining 10 double-roll papers are for pH ranges:
0.8 to 2.0, 1.3 to 4.4, 4.9 to 6.9, 6.0 to 8.0, 8.1 to 9.4, 1.4 to 2.8, 2.9 to 5.2, 5.5 to 8.0, 6.0 to 9.5, 9.2 to 10.6
Also included are a single roll and dispenser of Insta-Chek 0 to 13 and 3 rolls of Insta-Chek 0 to 13 refills.
Legal Information
Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
MicroFine is a trademark of Micro Essential Laboratory, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?