• USA Home
  • Z111953 - Demountable gas cell
EMAIL THIS PAGE TO A FRIEND
Z111953 Aldrich
Demountable gas cell
pathlength 50 mm
Purchase
Properties
Related Categories Labware, Sample Holders, Spectroscopy Supplies, Spectroscopy, IR
pathlength   50 mm
Description
General description
For IR analysis of pure gases or mixtures in which the components are present in concentrations of at least 5 to 10%. Cell is readily disassembled for cleaning. Includes Pyrex cell body with 2 stopcocks, Viton® O-rings, and PTFE gaskets. Requires two undrilled 32 mm diam. crystal windows (not included).
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
accessory
Product #
Description
Add to Cart
Z112100 32mm Round cell window, NaCl, Undrilled
Z112143 32mm Round cell window, Irtran-2, Undrilled
Z112135 32mm Round cell window, BaF2, Undrilled
Z267708 32mm Round cell window, KRS-5, Undrilled
Z112119 32mm Round cell window, KBr, Undrilled
Z267589 32mm Round cell window, CaF2, Undrilled
replacement part
Product #
Description
Add to Cart
Z111988 Replacement O-rings and gasket for demountable gas cell
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?