• USA Home
  • Z111961 - Demountable gas cell
EMAIL THIS PAGE TO A FRIEND
Z111961 Aldrich
Demountable gas cell
pathlength 100 mm
Purchase
Properties
pathlength   100 mm
Description
General description
For IR analysis of pure gases or mixtures in which the components are present in concentrations of at least 5 to 10%. Cell is readily disassembled for cleaning. Includes Pyrex cell body with 2 stopcocks, Viton® O-rings, and PTFE gaskets. Requires two undrilled 32 mm diam. crystal windows (not included).
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?