• USA Home
  • Z112321 - Presslok demountable cell holder
EMAIL THIS PAGE TO A FRIEND
Z112321 Aldrich
Presslok demountable cell holder
for window diameter, 32 mm
Purchase
Properties
Related Categories Labware, Sample Holders, Spectroscopy Supplies, Spectroscopy, IR
feature   for window diameter 32 mm
Description
General description
Assembly is accomplished by compressing cell plates between barrel assembly and self-locking cover. Highly efficient friction lock makes cell leak-proof. For use with all IR spectrometers. Pathlengths are varied by combinations of different PTFE spacers listed. Requires 25 or 32 mm diam. undrilled crystal windows listed (not included).
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?