• USA Home
  • Z112542 - Ohaus® mechanical balance
EMAIL THIS PAGE TO A FRIEND
Z112542 Aldrich
Ohaus® mechanical balance
Dial-O-Gram®
310
Purchase
Properties
feature   CE compliant
mfr. no.   Ohaus
measuring range   310 g weighing capacity
pan diam.   3.5 in.
Description
General description
• Die-cast aluminum beams
• Precision-ground steel knives
• Agate bearings
• SS pans
• Magnetic damping
Readings are made directly from large dial with vernier scale from 10 mg to 10 g. Values beyond 10 g are read through use of beams and riders. Includes specific gravity platform and double hook for specific gravity measurements. 0.01 g readability, 0.01 g sensitivity.
Legal Information
Dial-O-Gram is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?