• USA Home
  • Z112577 - Ohaus® mechanical balance
EMAIL THIS PAGE TO A FRIEND
Z112577 Aldrich
Ohaus® mechanical balance
750S triple-beam
Purchase
Properties
material   stainless steel weighing plate
feature   CE compliant
mfr. no.   Ohaus
measuring range   610 g weighing capacity
pan diam.   6 in.
Description
General description
• Die-cast aluminum beams
• Precision-ground steel knives
• Agate bearings
• SS pans
• Magnetic damping
Has three-tiered beams:
10 g in 0.1 g increments, 100 g in 10 g increments, and 500 g in 100 g increments. 0.1 g readability and 0.1 g sensitivity.
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?