• USA Home
  • Z112593 - Ohaus® mechanical balance
EMAIL THIS PAGE TO A FRIEND
Z112593 Aldrich
Ohaus® mechanical balance
Dial-O-Gram®
1650W
Purchase
Properties
material   stainless steel weighing plate
feature   CE compliant
mfr. no.   Ohaus
measuring range   610 g weighing capacity
pan diam.   6 in.
Description
General description
• Die-cast aluminum beams
• Precision-ground steel knives
• Agate bearings
• SS pans
• Magnetic damping
Similar to model 750S (Z112577), but has tare beam and direct-reading dial to 10 g in 0.1 g increments. 0.1 g readability, 0.1 g sensitivity.
Legal Information
Dial-O-Gram is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?