• USA Home
  • Z112771 - Ohaus® mechanical balance
EMAIL THIS PAGE TO A FRIEND
Z112771 Aldrich
Ohaus® mechanical balance
1119D Heavy-duty solution
Purchase
Properties
material   stainless steel weighing plate
feature   CE compliant
mfr. no.   Ohaus
measuring range   20,000 g weighing capacity
pan diam.   11 in.
Description
General description
• Die-cast aluminum beams
• Precision-ground steel knives
• Agate bearings
• SS pans
• Magnetic damping
Features undivided, extra-large capacity, 2.27 kg tare beam with locking poise. Two notched beams: 1,000 g in 100 g increments and 100 g in 1 g increments. 1 g readability, 1 g sensitivity. Five-piece, 20 kg weight set included. Has corrosion-resistant coating on exposed parts.
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?