• USA Home
  • Z113093 - Aldrich® glass reference electrode
EMAIL THIS PAGE TO A FRIEND
Z113093 Aldrich
Aldrich® glass reference electrode
General purpose, reference, Calomel
Purchase
Properties
Related Categories Aldrich, Aldrich pH Supplies, Glass reference electrode, Labware, Manufacturer Browser,
feature   flow rate moderate
  reference Calomel
  ceramic wick junction
parameter   0-80 °C temperature
Description
Application
pH, titrations
General description
The reference electrode is used as a pair with an indication electrode to make a measurement. All types supplied with a 30 in. lead. All reference half-cells have a universal reference pin connector.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?