• USA Home
  • Z113123 - Aldrich® glass reference electrode
EMAIL THIS PAGE TO A FRIEND
Z113123 Aldrich
Aldrich® glass reference electrode
Reverse sleeve, reference, Ag/AgCl
Purchase
Properties
Related Categories Aldrich, Aldrich pH Supplies, Glass reference electrode, Labware, Manufacturer Browser,
feature   flow rate high
  reference Ag/AgCl
  sleeve junction
parameter   −5-100 °C temperature
Description
Application
pH, ISE, emulsions, slurries, low ionic strength samples
General description
The reference electrode is used as a pair with an indication electrode to make a measurement. All types supplied with a 30 in. lead. All reference half-cells have a universal reference pin connector.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?