• USA Home
  • Z114464 - Wheaton soxhlet extraction apparatus
EMAIL THIS PAGE TO A FRIEND
Z114464 Aldrich
Wheaton soxhlet extraction apparatus
flask capacity 125 mL, top Joint: ST/NS 24/40, bottom Joint: ST/NS 34/45
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Extractors, Extractors, Labware, Manufacturer Browser,
Joint   bottom Joint: ST/NS34/45
  top Joint: ST/NS24/40
description   complete apparatus
mfr. no.   Wheaton Scientific
flask capacity   125 mL
Description
General description
Consists of extractor, Allihn condenser, and flask. Ground-glass joints on extractor and condenser.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?