• USA Home
  • Z114820 - Variable volume micropipette
EMAIL THIS PAGE TO A FRIEND
Z114820 Aldrich
Variable volume micropipette
volume 200-1000 μL
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettors, Sigma Pipettors More...
feature   subdivision 10 μL
mfr. no.   Wheaton Scientific
measuring range   1000 μL volume, precision: 0.3%
  200 μL volume, precision: 0.3%
volume   200-1000 μL
color   blue Fits microtips
Description
General description
Design of plunger stroke and overshoot movement allows tip to be flushed empty for maximum accuracy. Separate push buttons for volume delivery and tip-ejection. Color-coded plunger. Volumes are adjusted by rotating the plunger and reading them on the circular vernier. Order micropipette tips below.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
accessory
Product #
Description
Add to Cart
Z645508 Nichiryo pipette tips, volume range 200 μL, universal, bulk pack
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?