• USA Home
  • Z114898 - Wheaton positive displacement micropipette
EMAIL THIS PAGE TO A FRIEND
Z114898 Aldrich
Wheaton positive displacement micropipette
1-5 μL, accuracy: 1.0%
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Pipettors & Tips, Labware, Liquid Handling Equipment, Manufacturer Browser,
feature   subdivision μL 1,2,3,4,5
mfr. no.   Wheaton Scientific
measuring range   1-5 μL, accuracy: 1.0%
color   white capillary
Description
General description
Five preset volume stops. Graduated barrel slides and locks into any of the preset stops. Design eliminates errors and ensures consistent repetitive delivery. Sample drawn into capillary tube (listed below). No dead volume; constant volume regardless of viscosity. PTFE plunger tip.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?