EMAIL THIS PAGE TO A FRIEND
Z115290 Aldrich

Lab spoons

PTFE-coated, L 9 in.

Purchase

Properties

Related Categories Labware, Spatulas and Scoops More...
material   PTFE-coated
packaging   pkg of 1 ea
L   9 in.
spoon end size   1/2 in. × 1 in.

Description

General description

One spoon end, and one blade (5/16 × 2in.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?