• USA Home
  • Z118001 - Luer-to-Tubing Connector
EMAIL THIS PAGE TO A FRIEND
Z118001 Aldrich
Luer-to-Tubing Connector
male Luer lock to hose end, for tubing i.d., 1/83/16 in., nickel plated
Synonym: environmental, veterinary
Purchase
Properties
Related Categories Labware, Luer Lock Syringe Connectors, Syringe connectors, Syringes and Needles
material   nickel plated
feature   for tubing i.d. 1/83/16 in.
mfr. no.   Cadence Science, 6156
Description
General description
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating, easy to clean and resistant to corrosion.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?