• USA Home
  • Z118419 - Aldrich® heavy-base manometer
EMAIL THIS PAGE TO A FRIEND
Z118419 Aldrich
Aldrich® heavy-base manometer
0-120 torr pressure, (0 to 130 mbar)
Purchase
Description
General description
A constriction in the U-tube checks mercury surge if there is a sudden vacuum break. Scale is graduated in torr and mbar, and is movable for alignment with mercury level. Provided with a 2.5 mm glass stopcock. Filled with mercury.
How to use:
The manometer comes ready to operation, filled with 2 mL of mercury. Pull the grey round PVC bar out of the manometer tube and remove it before use. The manometer must be connected in between the vacuum pump and the process. To read the low pressure in (Millibar) push the zero marker on the sliding scale at the meniscus of the upgoing column. Measure directly at the opposite column.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
UN 2809 8/PG 3
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?