• USA Home
  • Z120138 - Satchel style cases
EMAIL THIS PAGE TO A FRIEND
Z120138 Aldrich
Satchel style cases
Single case
Purchase
Properties
Related Categories Cases, Containers, Labware More...
Description
General description
Gray co-polymer resin with hinged tongue-and-groove transparent lid. For storage of small parts, septa, bottle caps, corks, etc. 15 removable dividers with 9 to 24 adjustable compartments. 15 × 113/4 × 21/2 in.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?