• USA Home
  • Z120723 - Aldrich® spherical narrow-mouth Dewar flasks

EMAIL THIS PAGE TO A FRIEND
Z120723 Aldrich

Aldrich® spherical narrow-mouth Dewar flasks

capacity 2,000 mL, spherical, extended aluminum base

Purchase

Properties

Related Categories Aldrich, Aldrich Dewar Flasks & Accessories, Aldrich Spherical Narrow-Mouth Dewar Flasks, Flasks, Labware,
material   borosilicate glass
  narrow-mouth
sterility   non-sterile
packaging   pack of 1
mfr. no.   Sigma-Aldrich
Dewar I.D.   15.8 cm
capacity   2,000 mL
neck I.D.   3.2 cm

Description

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?