EMAIL THIS PAGE TO A FRIEND
Z120987 Aldrich
Eyewash fountain
Chrome-plated brass, pkg of 1 ea
Purchase
Properties
Related Categories Eye Protection Accessories, Eye Protection Equipment, Eyewash Equipment, Labware, Safety Equipment,
packaging   pkg of 1 ea
availability   not available in EU
Description
General description
Faucet-mounted eye and eye/face fountain mounts behind the spout, out of the way for everyday faucet use. Delivers over 3gpm water. Units will work with as little as 15lb of water pressure and as high as 50lb. Chrome-plated brass. Corresponding faucet adapters included. Meets ANSI Z358.1-1990.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?