• USA Home
  • Z121002 - Thermolyne® Test Tube Maxi-Mix II
EMAIL THIS PAGE TO A FRIEND
Z121002 Aldrich
Thermolyne® Test Tube Maxi-Mix II
AC input 240 V, 0.26 A
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Shakers, Labware, Manufacturer Browser, Shakers & Vortexers,
feature   CE compliant
mfr. no.   Barnstead International M37610-26
availability   not available in EU
Description
General description
For multi-tube or small flask mixing, use the 3 in. diam. rubber top or convert for single tube mixing by replacing pad with rubber cup. Three-position toggle switch for continuous or touch-control mixing (speed:100 to 3,000 rpm, no load). 6 × 41/2 × 7 in.
Legal Information
Maxi-Mix is a trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z645028 IKA® MS 3 basic shakers, universal small shaker, 230 V
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?