• USA Home
  • Z121401 - Hamilton® needles for RN syringes
EMAIL THIS PAGE TO A FRIEND
Z121401 Aldrich
Hamilton® needles for RN syringes
volume 2.5-100 μL, needle size 26s ga, bevel tip, needle L 101.5 mm (4 in.)
Purchase
Properties
Related Categories Analytical/Chromatography, Hamilton Needles, Hamilton Syringe Products, Hamilton needles, Hamilton syringes,
material   stainless steel needle (point style 2)
needle size   26s ga
feature   needle bevel tip
packaging   pkg of 6 ea
mfr. no.   Hamilton, 7804-04
needle L   101.5 mm (4 in.)
volume   2.5-100 μL
compatibility   for use with 2.5μL-100μL syringe
Description
Compatibility
For 5, 10 μL capacity
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?