• USA Home
  • Z122858 - Sigma-Aldrich® TLC Plates

EMAIL THIS PAGE TO A FRIEND
Z122858 Aldrich

Sigma-Aldrich® TLC Plates

cellulose matrix, L × W 20 cm × 20 cm

  •  eCl@ss 32110504

Purchase

Properties

Related Categories Analytical/Chromatography, Chromatography, Chromatography, TLC Plates, Glass TLC Plates: Cellulose matrix, LPLC/TLC/Paper Chromatography,
material   cellulose matrix
  glass support
feature   binder Proprietary
  fluorescent indicator
mfr. no.   Sigma-Aldrich
application(s)   thin layer chromatography (TLC): suitable
L × W   20 cm × 20 cm
layer thickness   250 μm
particle size   20 μm

Description

General description

Thin-layer chromatography (TLC) plates for adsorption, partition, and ion-exchange techniques. Six media choices with glass, polyester, and aluminum backings. Silica-based adsorbent layers in Sigma-Aldrich TLC plates incorporate a polymeric binder; highly purified silica gel plates incorporate a gypsum/polymer binder.
• Silica gel - for weakly to strongly polar separations, pore size 60 Å
• High purity silica gel - acid washed for separation of aflatoxins
• Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose)
• Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color.
• Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60Å

Application

Sigma-Aldrich® TLC Plates was used for TLC analysis during GTPase activity assay and lipid extraction/analysis.

Legal Information

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Pharma QC
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?