• USA Home
  • Z124230 - Normag liquid-liquid extractor
EMAIL THIS PAGE TO A FRIEND
Z124230 Aldrich
Normag liquid-liquid extractor
capacity 500 mL, for solvents lighter than water
Synonym: liquid liquid extraction, liquid liquid extractor, liquid-liquid extraction, liquid-liquid extractor
Purchase
Properties
Related Categories Analytical/Chromatography, Extraction Glassware, Extractors, Labware, Liquid / Liquid Extraction,
Joint   female Joint ST/NS top24/40
  male Joint ST/NS side arm24/40
mfr. no.   Normag
capacity   500 mL
Description
General description
A rotating distributor in the extractor vessel is driven by a magnetic stirring plate. The extractor solvent fed from the condenser to the distributor is centrifugally forced through the small holes in the distributor ring as fine droplets into the liquid to be extracted, producing optimum exchange of matter. The liquid to be extracted also rotates in the extractor.
The process is continuous and extraction is performed much more quickly than in traditional equipment. The rotary distributor on the inlet tube has a magnetic agitating rod, pivot nipple, bearings, antislip lock and separating ring with retaining ring.
Legal Information
Normag is a trademark of Normag Labor- und Prozesstechnik GmbH
Price and Availability
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?