EMAIL THIS PAGE TO A FRIEND
Z124729 Aldrich
Recessed septa
fits neck I.D., 17.0 mm
Purchase
Properties
Related Categories Labware, Septa, Serrated Septa More...
material   rubber septa (silicone)
feature   fits neck I.D. 17.0 mm
I.D.   ~10.6 mm
Description
General description
Made of silicone rubber, with recessed head. Can be used at high temperatures. Autoclavable.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?