• USA Home
  • Z125466 - Ace-Thred Thermometer/Tubing Adapter
EMAIL THIS PAGE TO A FRIEND
Z125466 Aldrich
Ace-Thred Thermometer/Tubing Adapter
Straight, Joint: ST/NS 24/40, thread I.D. 12.5-14.0 mm
Purchase
Properties
Related Categories Ace, Ace Adapters, Ace-Thred theremometer adapter and bushing, Glassware, Glassware Adapters,
label   A
Joint   Joint: ST/NS24/40
mfr. no.   Ace Glass 503040
thread I.D.   12.5-14.0 mm
Description
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replacement part
Product #
Description
Add to Cart
Z125393 Ace-Thred thermometer/tubing PTFE bushing, 7.5 mm hole
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?