• USA Home
  • Z126411 - Automatic shut-off quick-disconnect
EMAIL THIS PAGE TO A FRIEND
Z126411 Aldrich
Automatic shut-off quick-disconnect
coupling insert, bore 1/8 in., for tubing o.d., 0.25 in. (rigid)
Purchase
Properties
Related Categories CPC, Hose Coupling Tubing Connector, Labware, Manufacturer Browser, Tubing Connectors More...
feature   for tubing o.d. 0.25 in. (rigid)
mfr. no.   CRC Press
bore   1/8 in.
Description
General description
Spring-loaded, locking, made of PP with 316 SS springs and EPR seals. When pulled apart, both sides seal quickly and automatically, making couplings excellent for handling air-sensitive liquids. Available for flexible tubing or rigid PE and nylon tubing using a ferruleless nut. Max. pressure: 90 psi at 70°F. Working temp.: 32 to 230°F (air/water). Use same bore size for coupling insert and body.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?