• USA Home
  • Z126462 - Automatic shut-off quick-disconnect
EMAIL THIS PAGE TO A FRIEND
Z126462 Aldrich
Automatic shut-off quick-disconnect
coupling bodies, tube to pipe thread, bore 1/4 in., threaded joint, 3/8 in. NPT
Purchase
Properties
Related Categories CPC, Hose Coupling Tubing Connector, Labware, Manufacturer Browser, Tubing Connectors More...
feature   threaded joint 3/8 in. NPT
mfr. no.   CRC Press
bore   1/4 in.
Description
General description
Spring-loaded, locking, made of PP with 316 SS springs and EPR seals. When pulled apart, both sides seal quickly and automatically, making couplings excellent for handling air-sensitive liquids. Available for flexible tubing or rigid PE and nylon tubing using a ferruleless nut. Max. pressure: 90 psi at 70°F. Working temp.: 32 to 230°F (air/water). Use same bore size for coupling insert and body.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?