• USA Home
  • Z129917 - Thermolyne® fibrox-insulated heating tape
EMAIL THIS PAGE TO A FRIEND
Z129917 Aldrich
Thermolyne® fibrox-insulated heating tape
W × L 1/2 in. × 8 ft, output416 watts, Heavy-duty
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Heating Tapes, Fibrox-insulated, Heating,
AC input   120 V
mfr. no.   Barnstead International BIH051-080
availability   not available in EU
parameter   482 °C max. temp. (900 °F)
W × L   1/2 in. × 8 ft
output   416 watts
Description
General description
Serpentine configured heating elements embedded in tough, pliable, braided tape that conforms to almost any shape vessel for even, direct-contact heating without hot spots. All tapes have high temperature lead wires emerging from opposite ends and separable line plugs. A temperature controller is required.
Standard tape has finely stranded resistance wires doubly insulated with braided fibrox yarns and knitted into flat tape. Heavy-duty version has an extra braided fibrox covering over the standard tape, suitable for direct contact on conductive surfaces.
Legal Information
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?