• USA Home
  • Z130354 - Nalgene® FEP bottle

EMAIL THIS PAGE TO A FRIEND
Z130354 Aldrich

Nalgene® FEP bottle

Narrow-mouth, capacity 500 mL, opaque black Bottle

Synonym: fep bottles, nalgene bottles

Purchase

Properties

Related Categories Bottles and Caps, Fluorinated Plastic Bottles, Labware, Manufacturer Browser, Nalgene,
mfr. no.   Nalgene 1620
capacity   500 mL
color   black Screw closure
  opaque black Bottle

Description

General description

Maximum chemical and corrosion resistance at high and low temperatures. Excellent for trace metal analysis, storage of high-purity samples, and use with organic solvents. Autoclavable. Leakproof Tefzel® ETFE screw closures. Bottles with closures withstand temperatures from -105 to 150°C. Black opaque bottles are carbon black pigment and offers zero transmission of visible and UV light to 5,000 Å. Ideal storage for light sensitive materials.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?