• USA Home
  • Z130559 - Nalgene® narrow-mouth bottles
EMAIL THIS PAGE TO A FRIEND
Z130559 Aldrich
Nalgene® narrow-mouth bottles
size 60 mL
Synonym: HDPE bottles, nalgene bottle, nalgene bottles
Purchase
Properties
Related Categories Amber High-Density Polyethylene Bottles, Bottles and Caps, Labware, Manufacturer Browser, Nalgene,
material   amber high-density polyethylene bottle
packaging   pkg of 12 ea
mfr. no.   Nalgene® 2004
size   60 mL
Description
General description
Popular all-purpose containers for use in most laboratory applications. Not autoclavable. Protects light-sensitive liquids.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?