• USA Home
  • Z131326 - Nalgene® polypropylene Erlenmeyer flask
EMAIL THIS PAGE TO A FRIEND
Z131326 Aldrich
Nalgene® polypropylene Erlenmeyer flask
capacity 50 mL
Synonym: conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
Purchase
Properties
Related Categories Erlenmeyer Flasks, Plastic, Flasks, Labware, Manufacturer Browser, Nalgene,
material   polypropylene
sterility   non-sterile
feature   cap: No
packaging   pkg of 12 ea
mfr. no.   Nalgene® 4102
capacity   50 mL
stopper size size   1
Description
General description
Autoclavable,
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?