• USA Home
  • Z131873 - Nalgene® PolyPaper labels
EMAIL THIS PAGE TO A FRIEND
Z131873 Aldrich
Nalgene® PolyPaper labels
size 22 mm ( 7/8 in.) × 22 mm ( 7/8 in.)
Purchase
Properties
Related Categories General Labels, Labels and Markers, Labels for Laser Printers, Labware, Manufacturer Browser,
packaging   pkg of 600 ea
mfr. no.   Nalgene
size   22 mm ( 7/8 in.) × 22 mm ( 7/8 in.)
Description
General description
Coated and die-cut with rounded corners. Special pressure-sensitive adhesive adheres well to plastics, glass, or metals. Not for use in photocopiers or laser printers.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
PolyPaper is a trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?