• USA Home
  • Z131938 - Nalgene® PolyPaper labels
EMAIL THIS PAGE TO A FRIEND
Z131938 Aldrich
Nalgene® PolyPaper labels
size 51 mm (2 in.) × 102 mm (4 in.)
Purchase
Properties
Related Categories General Labels, Labels and Markers, Labels for Laser Printers, Labware, Manufacturer Browser,
packaging   pkg of 60 ea
mfr. no.   Nalgene
size   51 mm (2 in.) × 102 mm (4 in.)
Description
General description
Coated and die-cut with rounded corners. Special pressure-sensitive adhesive adheres well to plastics, glass, or metals. Not for use in photocopiers or laser printers.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
PolyPaper is a trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?