• USA Home
  • Z133728 - Whatman® cellulose extraction thimbles
EMAIL THIS PAGE TO A FRIEND
Z133728 Aldrich
Whatman® cellulose extraction thimbles
I.D. × L 19 mm × 90 mm
Purchase
Properties
Related Categories Extraction Thimbles, Extractors, Labware More...
packaging   pkg of 25 ea
mfr. no.   Whatman
I.D. × L   19 mm × 90 mm
Description
General description
Whatman® Cellulose Extraction Thimbles
High Performance Cellulose Extraction Thimbles
Cellulose extraction thimbles are produced from high-quality alpha cellulose cotton linter and have excellent mechanical strength and retention.
Standard single thickness thimbles have a wall thickness of approximately 1 mm (10.0 μm nominal particle retention).
Double thickness thimbles have a wall thickness of approximately 2 mm (6.0 μm nominal particle retention) for applications where higher retention and increased wet or dry strength, or rigidity are required.
The high purity of the materials ensures reliable and reproducible analytical results.
Standard Cellulose Extraction Thimbles
Thimbles of type 603 are made from high-quality cellulose and 603 g thimbles are made from borosilicate glass fibers with an inorganic binder. For all automated extraction apparatus in common use, Whatman offers thimbles whose dimensions are matched exactly to those of the thimble holders to ensure optimal fit. Dimensions shown are for internal diameter and external length.
Legal Information
Whatman is a registered trademark of GE Healthcare
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?