Z134147 Aldrich

Whatman® Panpeha pH indicator strips

pH range 0 to 14, 200/pk

DOWNLOAD MSDS (PDF)

Synonym: pH indicator papers, pH indicator strips, pH papers, pH strips

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

General description

Whatman® Universal Indicator Papers

Whatman universal indicator papers have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular color. A check against the color comparision table supplied allows the pH to be determined

PANPEHA Nr. 112
Measure pH values from 0 to 14 in 24 graduations. Sensitivity of 0.5 pH unit in the 0 to 9 range; 1 pH unit in the 9 to 14 range. Box of 200 strips with color reference chart.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Panpeha is a trademark of Sigma-Aldrich Co. LLC

Whatman is a registered trademark of GE Healthcare

Price and Availability

Customers Also Viewed

Hydrion<SUP>®</SUP> Brilliant pH dip stiks

pH-range 1 - 14

pH test strips

0-14 pH, resolution: 1.0 pH unit

pH test strips

4.5-10.0 pH, resolution: 0.5 pH unit

Triple color chart

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?