• USA Home
  • Z135887 - Write-on label tape
EMAIL THIS PAGE TO A FRIEND
Z135887 Aldrich
Write-on label tape
W 1/2 in.
Purchase
Properties
Related Categories Bel-Art, Bel-Art Labels & Markers, General Labels, Label Tape, Labels and Markers,
mfr. no.   Bel-Art
W   1/2 in.
Description
General description
For applying labels to surfaces of any contour or material (glass, metal, plastic). Requires no special stylus: just pencil, ballpoint pen, or solvent ink marker. Resists moisture and most solvents, and can be used at temperatures of -73 to 121 °C. Will withstand repeated autoclaving and remain legible under most lab use. May be readily applied and peeled off without leaving sticky residue. 40 yd roll.
Legal Information
Write-on is a trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?