• USA Home
  • Z136506 - Agate mortar and pestle
EMAIL THIS PAGE TO A FRIEND
Z136506 Aldrich
Agate mortar and pestle
Standard form, O.D. 125 mm
Purchase
Properties
Related Categories Labware, Mortar and Pestle More...
material   agate
D   30 mm
I.D.   110 mm
O.D.   125 mm
Description
General description
Made of natural agate with highly polished grinding surfaces. Excellent for preparing laboratory samples and high purity powders. The agate material is a special silicate with a main composition of SiO2. It is resistant to detergent. The thermal endurance is limited up to 350°C.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?