• USA Home
  • Z136794 - Flex-Grip® stirrer chuck
EMAIL THIS PAGE TO A FRIEND
Z136794 Aldrich
Flex-Grip® stirrer chuck
fits shaft o.d., 19 mm
Purchase
Description
General description
Nylon chuck for use with stirring shafts. Has flexible insert that allows for misalignment of shaft without danger of breakage. Fits 8mm motor shaft.
Legal Information
Flex-Grip is a registered trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?