• USA Home
  • Z136905 - Firestone rotary evaporators
EMAIL THIS PAGE TO A FRIEND
Z136905 Aldrich
Firestone rotary evaporators
for flask size, ST/NS: 24/40
Purchase
Properties
Related Categories Ace, Ace Adapters, Ace Firestone Rotary Evaporators, Evaporators, Firestone Evaporators & Accessories,
feature   for flask size ST/NS: 24/40
mfr. no.   Ace Glass 671440
Description
General description
Constructed of glass, PTFE, and nylon. Operates without metal devices. Hollow glass tube (10 mm o.d.), with joint at one end for flask and holes drilled near center for vacuum. Attaches to any laboratory stirring motor. Tube turns inside PTFE bearing, threads held in by nylon bushing, saddle O-ring, and lock nut. 2 mm straight-bore stopcock attaches to drying tube or McLeod gauge. One arm of 4 mm bore double stopcock attaches to dry ice trap and vacuum line; other arm is for easy vacuum release and can be connected to a trap. Complete set includes joint clamp.
NOTE: Designed by Dr. Raymond Firestone, Merck, Sharp & Dohme Research Labs.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?