• USA Home
  • Z137030 - Ace dry ice twin chamber condenser-trap
EMAIL THIS PAGE TO A FRIEND
Z137030 Aldrich
Ace dry ice twin chamber condenser-trap
#50 bushing only
Purchase
Description
General description
Includes twin chamber, two dry ice finger condensers, and two nylon bushings with FETFE® O-rings. Overall height of 18 in. Finger condensers secured in chambers via #50 Ace-Threds using bushings and O-rings, allowing for easier removal for cleaning. 3/8 in. O.D.hose ports. 0 to 5 mm easy action threaded style stopcock with double PTFE ring seals on plug for draining both chambers simultaneously. Dry ice fingers have 450 mL capacity each. The chambers have 600 mL capacity when fingers are inserted.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?