• USA Home
  • Z137413 - Firestone Hy-N-Dry desiccator stopper
EMAIL THIS PAGE TO A FRIEND
Z137413 Aldrich
Firestone Hy-N-Dry desiccator stopper
Joint: ST/NS 24/40
Purchase
Properties
Related Categories Ace, Ace Stoppers, Glass Stoppers, Labware, Manufacturer Browser,
Joint   Joint: ST/NS24/40
mfr. no.   Ace Glass 827719
Description
General description
Sintered-glass disc (porosity: 70 to 100 μm) sealed into bottom of stopper. When filled with 10 to 20 mesh drying agents, covered with plastic cap, and inserted into any ST joint vessel, becomes an inexpensive desiccator. Pinhole in cap allows assembled unit to "breathe" through, not around, desiccant with temperature fluctuations. Filled with Drierite®, ST 24/40 stopper will absorb up to 1g of water. Plastic cap included.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?