EMAIL THIS PAGE TO A FRIEND
Z146307 Aldrich
Sampler bottle kit
no. of bottles, 24, bottle capacity 6.0 mL
Purchase
Properties
Related Categories Aldrich, Aldrich Vials & Caps, Labware, Manufacturer Browser
feature   no. of bottles 24
bottle capacity   6.0 mL
Description
General description
Designed for safe, convenient shelf storage of laboratory samples. Each Sampler includes Aldrich Kitcase, Wheaton Shorty® Vials with poly-foam-lined screw-caps, and labels for bottles, Kitcase spine, and index.
Legal Information
Shorty Vial is a registered trademark of Wheaton Industries, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?