• USA Home
  • Z146536 - Ace Rotary-evaporator/freeze-drying flask
EMAIL THIS PAGE TO A FRIEND
Z146536 Aldrich
Ace Rotary-evaporator/freeze-drying flask
capacity 1,000 mL
Purchase
Properties
Related Categories Ace, Ace Flasks, Ace Rotary-Evaporator/Feeze-Drying Flasks, Evaporating Flasks, Evaporators,
mfr. no.   Ace Glass 703008
capacity   1,000 mL
Description
General description
Flange with two ground flats opposite each other allows nylon retainers on adapter head to pass over. O-ring in head and flask grooves make a vacuum-tight seal when head is turned 90° in either direction. No grease, springs, or clamp necessary. Adapter heads and O-rings sold separately.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?