• USA Home
  • Z148156 - Storage bottles with septum-inlet
EMAIL THIS PAGE TO A FRIEND
Z148156 Aldrich
Storage bottles with septum-inlet
capacity 250 mL, glass stopcock
Purchase
Properties
Related Categories Air-Sensitive Chemistry, Aldrich Air Sensitive, Aldrich Air Sensitive Storage Bottles, Bottles and Flasks, Dispensing and Storage,
material   clear glass
  glass stopcock
Joint   Joint ST/NS24/40 threaded
capacity   250 mL
Description
General description
All septum inlets have 4 mm bore stopcocks requiring rubber septa, Z553913 or Z554030.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?