• USA Home
  • Z148768 - Uvex® Futura 9300 goggles
EMAIL THIS PAGE TO A FRIEND
Z148768 Aldrich
Uvex® Futura 9300 goggles
with wide curve, antifog PC lens for unrestricted field of view
Purchase
Properties
Related Categories Eye Protection Equipment, Goggles, Labware, Safety Equipment
availability   not available in EU
Description
General description
With wide curve, antifog PC lens for unrestricted field of view. Unique ventilation system keeps out liquids, flying particles, and dust. 95% UV protection from 200-385 nm. Meets ANSI Z87.1-1989 and CSAZ94.3-1988 requirements.
Legal Information
Futura is a trademark of Magid Glove & Safety Mfg. Co.
Uvex is a registered trademark of Uvex Winter Optik GmbH Ltd.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?