EMAIL THIS PAGE TO A FRIEND
Z150444 Aldrich
Keck® Joint Clip
standard taper, joint size, 24, green
Purchase
Properties
Related Categories Chemistry Equipment and Labware, Clamps and Supports, Joint Clamps, Labware
feature   joint size 24
color   green
Description
General description
Color coded, Delrin® plastic joint clips will not scratch glassware or corrode. Withstand temperatures up to 302 °F (150 °C). Resistant to concentrated alkalis and diluted acids; not resistant to halogen gases or concentrated acids.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?