EMAIL THIS PAGE TO A FRIEND
Z151998 Aldrich
Organic Reactions
Volume 4
  •  ISBN-10 0471005614
  •  ISBN-13 978-047100561-2
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   R.J. Adams, John Wiley & Sons, Ltd, 1948, 428 pp., hard cover
mfr. no.   Wiley
Description
General description
Presents critical discussions of widely used organic reactions from a preparative view point.
The volumes of Organic Reactions are collections of chapters each devoted to a single reaction, or a definite phase of a reaction, of wide applicability. The material is treated from a preparative viewpoint, with emphasis on limitations, interfering influences, effects of structure, and the selection of experimental techniques. Numerous detailed procedures illustrate the significant modifications of each method. Includes tables that contain all possible examples of the reaction under consideration.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?