• USA Home
  • Z152366 - Aldrich® manostats
EMAIL THIS PAGE TO A FRIEND
Z152366 Aldrich
Aldrich® manostats
porosity 10-20 μm
Purchase
Description
General description
Precisely regulates pressure between vacuum source and the distillation system for gradients in pressure less than 100mm Hg. Requires approx. 20 mL of mercury, not included. See Technical Information Bulletin No. AL-153 for instructions.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?