• USA Home
  • Z152382 - Stainless steel 304 syringe needle, noncoring point
EMAIL THIS PAGE TO A FRIEND
Z152382 Aldrich
Stainless steel 304 syringe needle, noncoring point
gauge 12, L 36 in.
Purchase
Properties
Related Categories Aldrich, Aldrich Syringes & Needles, Labware, Manufacturer Browser, Non-Coring,
description   Standard Hub with 304 Stainless Steel Tubing
packaging   pack of 1 ea
mfr. no.   Cadence Science
L   36 in.
gauge   12
Description
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Syringe Needle Gauge Chart
From our library of Related Content, Sigma-Aldrich presents Syringe Needle Gauge Chart
Related Products
comparable product
Product #
Description
Add to Cart
C0918 Fluorimeter cuvettes, Polystyrene, without stopper
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?