• USA Home
  • Z152390 - Stainless steel 304 syringe needle, noncoring point
EMAIL THIS PAGE TO A FRIEND
Z152390 Aldrich
Stainless steel 304 syringe needle, noncoring point
gauge 14, L 36 in.
Purchase
Properties
Related Categories Aldrich, Aldrich Syringes & Needles, Labware, Manufacturer Browser, Non-Coring,
description   Standard Hub with 304 Stainless Steel Tubing
packaging   pack of 1 ea
mfr. no.   Cadence Science
L   36 in.
gauge   14
Description
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?